missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dichlorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. D29271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29271G 1 g
D29275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29271G Supplier TCI America Supplier No. D29271G

Chemical Identifiers

CAS 21581-45-3
Molecular Formula C8H10Cl2N
Molecular Weight (g/mol) 191.07
MDL Number MFCD00060617
InChI Key MQPUAVYKVIHUJP-UHFFFAOYSA-O
Synonym 3,4-dichlorophenethylamine, 2-3,4-dichlorophenyl ethanamine, 2-3,4-dichlorophenyl ethylamine, 2-3,4-dichlorophenyl ethan-1-amine, 2-3,4-dichloro-phenyl-ethylamine, benzeneethanamine, 3,4-dichloro, 2-3,4,-dichlorophenyl ethylamine, 1-amino-2-3,4-dichlorophenyl ethane, pubchem12521, acmc-1apis
PubChem CID 217958
IUPAC Name 2-(3,4-dichlorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=C(Cl)C(Cl)=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 190.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(3,4-Dichlorophenyl)ethylamine