missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dichlorophenyl)ethanol 96.0+%, TCI America™

Catalog No. D29235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29235G Supplier TCI America Supplier No. D29235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35364-79-5
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.051
MDL Number MFCD00800673
InChI Key GITOMJDYNUMCOV-UHFFFAOYSA-N
Synonym 2-3,4-dichlorophenyl ethanol, 3,4-dichlorophenethyl alcohol, 3,4-dichlorophenethylalcohol, 2-3,4-dichlorophenyl ethan-1-ol, benzeneethanol, 3,4-dichloro, 2-3,4-dichlorophenyl-ethanol, 3,4-dichlorobenzeneethanol, benzeneethanol,3,4-dichloro, acmc-1adlk, 2-3,4-dichloro-phenyl-ethanol
PubChem CID 244558
IUPAC Name 2-(3,4-dichlorophenyl)ethanol
SMILES C1=CC(=C(C=C1CCO)Cl)Cl

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 g
Formula Weight 191.05
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3,4-Dichlorophenyl)ethanol