missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetrachlorophenol Sodium Salt 89.0+%, TCI America™

Catalog No. T006925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T006925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T006925G Supplier TCI America Supplier No. T006925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131-61-3
Molecular Formula C6HCl4NaO
Molecular Weight (g/mol) 253.86
MDL Number MFCD00019980
InChI Key YLFFQZKUOUYUFG-UHFFFAOYSA-M
PubChem CID 23616151
IUPAC Name sodium 2,3,4,6-tetrachlorobenzen-1-olate
SMILES [Na+].[O-]C1=C(Cl)C(Cl)=C(Cl)C=C1Cl

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 253.86
Percent Purity ≥89.0% (GC)
Chemical Name or Material 2,3,4,6-Tetrachlorophenol Sodium Salt