missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzyl-beta-D-glucopyranosyl Fluoride 96.0+%, TCI America™

Catalog No. t1923500mg
Change view
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
T1923500MG 500 mg
1 options

Catalog No. T1923500MG

Supplier: TCI America T1923500MG

Only null left

Chemical Identifiers

CAS 78153-79-4
Molecular Formula C34H35FO5
Molecular Weight (g/mol) 542.65
MDL Number MFCD01862264
InChI Key QNXIKNZDQVSBCO-UHFFFAOYNA-N
PubChem CID 10886059
IUPAC Name 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-fluorooxane
SMILES FC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Specifications

Melting Point 47°C
Color White
Quantity 500 mg
Formula Weight 542.65
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-beta-D-glucopyranosyl Fluoride
TSCA No
Recommended Storage Refrigerator