missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl Fluoride 95.0+%, TCI America™

Catalog No. T1922500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1922500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1922500MG Supplier TCI America Supplier No. T1922500MG
Only null left

Chemical Identifiers

CAS 89025-46-7
Molecular Formula C34H35FO5
Molecular Weight (g/mol) 542.65
MDL Number MFCD13195515
InChI Key QNXIKNZDQVSBCO-UHFFFAOYNA-N
PubChem CID 11092783
IUPAC Name 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-fluorooxane
SMILES FC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Specifications

Melting Point 69°C
Color White
Quantity 500 mg
Formula Weight 542.65
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl Fluoride
TSCA No
Recommended Storage Refrigerator