missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzoyl-D-mannopyranose 93.0+%, TCI America™

Catalog No. T20561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20561G 1 g
T20565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20561G Supplier TCI America Supplier No. T20561G
Only null left

Chemical Identifiers

CAS 113544-59-5
Molecular Formula C34H28O10
Molecular Weight (g/mol) 596.59
MDL Number MFCD02683399
InChI Key FCDYAJBVISGNLC-UHFFFAOYNA-N
PubChem CID 11146396
IUPAC Name [3,4,5-tris(benzoyloxy)-6-hydroxyoxan-2-yl]methyl benzoate
SMILES OC1OC(COC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 1 g
Formula Weight 596.59
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzoyl-D-mannopyranose
TSCA No
Recommended Storage Freezer