Learn More
Thermo Scientific Chemicals 2,3,4,6-Tetra-O-acetyl-D-glucopyranose, 98%
CAS: 10343-06-3 | C14H20O10 | 348.304 g/mol
Supplier: Thermo Scientific Chemicals J6605206
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform or ethyl acetate
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 2,3,4,6-Tetra-O-acetyl-D-glucopyranose | |
| 137°C to 138°C | |
| C14H20O10 | |
| 5 g | |
| Soluble in chloroform or ethyl acetate | |
| CC(=O)OCC1C(C(C(C(O1)O)OC(=O)C)OC(=O)C)OC(=O)C | |
| 348.304 | |
| 348.3 | |
| Powder |
| 10343-06-3 | |
| White | |
| MFCD00061634 | |
| 2,3,4,6-tetra-o-acetyl-d-glucopyranose, 2,3,4,6-tetra-o-acetylglucopyranose, 2r,3r,4s,5r-3,4,5-tris acetyloxy-6-hydroxyoxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-beta-d-glucopyranose, pubchem10600, d-glucopyranose 2,3,4,6-tetraacetate, 2,3,4,6-tetra-o-acetyl-glucopyranose, 2,3,4,6-tetraacetate d-glucopyranose, d-glucopyranose, 2,3,4,6-tetraacetate, 2r,3r,4s,5r-2-acetoxymethyl-6-hydroxytetrahydro-2h-pyran-3,4,5-triyl triacetate | |
| IEOLRPPTIGNUNP-RQICVUQASA-N | |
| [(2R,3R,4S,5R)-3,4,5-triacetyloxy-6-hydroxyoxan-2-yl]methyl acetate | |
| 11824227 | |
| 98% |
Chemical Identifiers
| 10343-06-3 | |
| 348.304 | |
| IEOLRPPTIGNUNP-RQICVUQASA-N | |
| 11824227 | |
| CC(=O)OCC1C(C(C(C(O1)O)OC(=O)C)OC(=O)C)OC(=O)C |
| C14H20O10 | |
| MFCD00061634 | |
| 2,3,4,6-tetra-o-acetyl-d-glucopyranose, 2,3,4,6-tetra-o-acetylglucopyranose, 2r,3r,4s,5r-3,4,5-tris acetyloxy-6-hydroxyoxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-beta-d-glucopyranose, pubchem10600, d-glucopyranose 2,3,4,6-tetraacetate, 2,3,4,6-tetra-o-acetyl-glucopyranose, 2,3,4,6-tetraacetate d-glucopyranose, d-glucopyranose, 2,3,4,6-tetraacetate, 2r,3r,4s,5r-2-acetoxymethyl-6-hydroxytetrahydro-2h-pyran-3,4,5-triyl triacetate | |
| [(2R,3R,4S,5R)-3,4,5-triacetyloxy-6-hydroxyoxan-2-yl]methyl acetate |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only