missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3,4,6-Tetra-O-acetyl-D-glucopyranose, 98%

Catalog No. AAJ6605203
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6605203 1 g
AAJ6605206 5 g
AAJ6605209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6605203 Supplier Thermo Scientific Chemicals Supplier No. J6605203

CAS: 10343-06-3 | C14H20O10 | 348.304 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform or ethyl acetate

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 10343-06-3
Molecular Formula C14H20O10
Molecular Weight (g/mol) 348.304
MDL Number MFCD00061634
InChI Key IEOLRPPTIGNUNP-RQICVUQASA-N
Synonym 2,3,4,6-tetra-o-acetyl-d-glucopyranose, 2,3,4,6-tetra-o-acetylglucopyranose, 2r,3r,4s,5r-3,4,5-tris acetyloxy-6-hydroxyoxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-beta-d-glucopyranose, pubchem10600, d-glucopyranose 2,3,4,6-tetraacetate, 2,3,4,6-tetra-o-acetyl-glucopyranose, 2,3,4,6-tetraacetate d-glucopyranose, d-glucopyranose, 2,3,4,6-tetraacetate, 2r,3r,4s,5r-2-acetoxymethyl-6-hydroxytetrahydro-2h-pyran-3,4,5-triyl triacetate
PubChem CID 11824227
IUPAC Name [(2R,3R,4S,5R)-3,4,5-triacetyloxy-6-hydroxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)O)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 137°C to 138°C
Color White
Quantity 1 g
Solubility Information Soluble in chloroform or ethyl acetate
Formula Weight 348.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-D-glucopyranose
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only