missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl 2,2,2-Trichloroacetimidate 98.0+%, TCI America™

Catalog No. p-7137242
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24911G 1 g
T24915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24911G Supplier TCI America Supplier No. T24911G

Chemical Identifiers

CAS 92052-29-4
Molecular Formula C16H20Cl3NO10
Molecular Weight (g/mol) 492.683
InChI Key IBUZGVQIKARDAF-RGDJUOJXSA-N
PubChem CID 11038322
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-(2,2,2-trichloroethanimidoyl)oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=N)C(Cl)(Cl)Cl)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 158°C
Color White
Quantity 1 g
Formula Weight 492.68
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Freezer