Learn More
2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%, Thermo Scientific™
Chemical Identifiers
| CAS | 572-09-8 |
|---|---|
| Molecular Formula | C14H19BrO9 |
| Molecular Weight (g/mol) | 411.2 |
| MDL Number | MFCD00063254 |
| InChI Key | CYAYKKUWALRRPA-RGDJUOJXSA-N |
| Synonym | 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromglucose, acetobromoglucose, a-acetobromglucose, a-acetobromglucose, 2,3,4,6-tetra-o-acetyl-, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate |
| PubChem CID | 101776 |
| IUPAC Name | [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate |
| SMILES | CC(=O)OCC1C(C(C(C(O1)Br)OC(=O)C)OC(=O)C)OC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC270930100
|
Thermo Scientific Chemicals
270930100 |
10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC270930500
|
Thermo Scientific Chemicals
270930500 |
50g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 572-09-8 | |
| 411.2 | |
| CYAYKKUWALRRPA-RGDJUOJXSA-N | |
| 101776 | |
| CC(=O)OCC1C(C(C(C(O1)Br)OC(=O)C)OC(=O)C)OC(=O)C |
| C14H19BrO9 | |
| MFCD00063254 | |
| 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromglucose, acetobromoglucose, a-acetobromglucose, a-acetobromglucose, 2,3,4,6-tetra-o-acetyl-, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate | |
| [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate |
Specifications
| 572-09-8 | |
| 100.0 | |
| 98% min. (Argentometry) | |
| C14H19BrO9 | |
| 10g | |
| +194.5° (20°C c=2,chloroform) | |
| 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromglucose, acetobromoglucose, a-acetobromglucose, a-acetobromglucose, 2,3,4,6-tetra-o-acetyl-, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate | |
| CYAYKKUWALRRPA-RGDJUOJXSA-N | |
| [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate | |
| 2% CaCO3 | |
| 411.2 | |
| 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Stabilized |
| 98.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00063254 | |
| 15, 59 | |
| + 194.50 | |
| Solubility in water: decomposes. Other solubilities: 1g/20mL abs. ethanol - more soluble in methanol, freely soluble in ether, chloroform, acetone,, ethyl acetate and benzene, slightly soluble in petroleum ether | |
| CC(=O)OCC1C(C(C(C(O1)Br)OC(=O)C)OC(=O)C)OC(=O)C | |
| 411.2 | |
| 101776 | |
| 98.5% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 209-339-
RUO â Research Use Only