missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%

Catalog No. AC270930500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270930500 50 g Glass bottle
AC270930100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270930500 Supplier Thermo Scientific Chemicals Supplier No. 270930500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 572-09-8 | C14H19BrO9 | 411.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 572-09-8
Molecular Formula C14H19BrO9
Molecular Weight (g/mol) 411.20
MDL Number MFCD00063254
InChI Key CYAYKKUWALRRPA-HAGGZJDPNA-N
Synonym 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, acetobromo glucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate
PubChem CID 101776
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 83.0°C to 88.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass bottle
Quantity 50 g
Merck Index 15, 59
Specific Rotation Condition +194.5° (20°C c=2,chloroform)
Specific Rotation + 194.50
Solubility Information Solubility in water: decomposes. Other solubilities: 1g/20mL abs. ethanol - more soluble in methanol, freely soluble in ether, chloroform, acetone,, ethyl acetate and benzene, slightly soluble in petroleum ether
Stabilizer 2% CaCO3
Formula Weight 411.2
Percent Purity 98.5%
Physical Form Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 209-339-

RUO – Research Use Only