missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl 2,2,2-Trichloroacetimidate 95.0+%, TCI America™

Catalog No. T22955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22955G 5 g
T22951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22955G Supplier TCI America Supplier No. T22955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86520-63-0 | C16H20Cl3NO10 | 492.683 g/mol

Chemical Identifiers

CAS 86520-63-0
Molecular Formula C16H20Cl3NO10
Molecular Weight (g/mol) 492.683
MDL Number MFCD07369652
InChI Key IBUZGVQIKARDAF-MBJXGIAVSA-N
PubChem CID 11092215
IUPAC Name [(2R,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-(2,2,2-trichloroethanimidoyl)oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=N)C(Cl)(Cl)Cl)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 122°C
Color Yellow
Quantity 5 g
Formula Weight 492.68
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Freezer