missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%

CAS: 14131-84-1 | C12H20O6 | 260.286 g/mol

Supplier:  Thermo Scientific Chemicals J6630314

 View more versions of this product

Catalog No. AAJ6630314

Only null left
Explore available promotions

Description

Description

2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

Solubility
Soluble in acetone, methanol, water.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
Specifications

Specifications

2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose
125°C to 126°C (decomposition)
C12H20O6
25 g
Hygroscopic
Soluble in acetone,methanol,water.
CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C
260.286
260.28
Powder
14131-84-1
White
MFCD00134206
84382
2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
JWWCLCNPTZHVLF-WVHJSVDISA-N
(3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
45039045
98%

Chemical Identifiers

14131-84-1
260.286
JWWCLCNPTZHVLF-WVHJSVDISA-N
45039045
CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C
C12H20O6
MFCD00134206
2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
(3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only