missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%

Catalog No. AAJ6630306
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6630306 5 g
AAJ6630309 10 g
AAJ6630314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6630306 Supplier Thermo Scientific Chemicals Supplier No. J6630306

CAS: 14131-84-1 | C12H20O6 | 260.286 g/mol

2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

Solubility
Soluble in acetone, methanol, water.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.

Chemical Identifiers

CAS 14131-84-1
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00134206
InChI Key JWWCLCNPTZHVLF-WVHJSVDISA-N
Synonym 2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
PubChem CID 45039045
IUPAC Name (3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
SMILES CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C

Specifications

Melting Point 125°C to 126°C (decomposition)
Color White
Quantity 5 g
Beilstein 84382
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,methanol,water.
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only