missing translation for 'onlineSavingsMsg'
Learn More

(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic Acid Monohydrate 97.0+%, TCI America™

Catalog No. d21915g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21915G Supplier TCI America Supplier No. D21915G

Chemical Identifiers

CAS 68539-16-2
Molecular Formula C12H20O8
Molecular Weight (g/mol) 292.284
MDL Number MFCD00150517
InChI Key ZFQRGFMVXLSLKZ-QCILGFJPSA-N
Synonym (-)-2,3:4,6-Di-O-isopropylidene-alpha-L-xylo-2-hexulofuranosonic Acid, Dikegulac
PubChem CID 16211407
IUPAC Name (3aR,4aS,8aR,8bS)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine-3a-carboxylic acid;hydrate
SMILES CC1(OCC2C(O1)C3C(O2)(OC(O3)(C)C)C(=O)O)C.O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 274.27
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic Acid Monohydrate
TSCA No
Recommended Storage Refrigerator