missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine, 95%

Catalog No. AAH27843MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27843MD 250 mg
AAH2784303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27843MD Supplier Thermo Scientific Chemicals Supplier No. H27843MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5946-39-4 | C9H12N2 | 148.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5946-39-4
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD03789577
InChI Key MLXBHOCKBUILHN-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepine
PubChem CID 2771762
IUPAC Name 2,3,4,5-tetrahydro-1H-1,4-benzodiazepine
SMILES C1CNC2=CC=CC=C2CN1

Specifications

Melting Point 96°C to 100°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 148.21
Percent Purity 95%
Chemical Name or Material 2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only