missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%

Catalog No. p-7043248
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6653709 10 g
AAJ6653718 50 g
AAJ6653722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6653709 Supplier Thermo Scientific Chemicals Supplier No. J6653709
Only null left

CAS: 20880-92-6 | C12H20O6 | 260.286 g/mol

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

Solubility
Soluble in chloroform, ethyl acetate, methanol.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 20880-92-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00022183
InChI Key PSSHGMIAIUYOJF-OZFQHSNDSA-N
Synonym 2,3:4,5-bis-o-1,2 methylethylidene-beta-d-fruetopyranose
PubChem CID 45357248
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
SMILES CC1(OC2COC3(C(C2O1)OC(O3)(C)C)CO)C

Specifications

Melting Point 93°C to 96°C
Color White
Quantity 10 g
Solubility Information Soluble in chloroform,ethyl acetate,methanol.
Optical Rotation −25.5° (c=1 in Chloroform)
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only