missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethylindolenine 97.0+%, TCI America™

Catalog No. T076625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T076625ML 25 mL
T0766500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T076625ML Supplier TCI America Supplier No. T076625ML

Chemical Identifiers

CAS 1640-39-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00005724
InChI Key FLHJIAFUWHPJRT-UHFFFAOYSA-N
Synonym 2,3,3-trimethylindolenine, 2,3,3-trimethyl-3h-indole, 3h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin, 2,3,3-trimethyl-3-hydroindole, pubchem7445, rarechem ah bs 0130, 2,3-trimethylindolenine, 2,3,3-trimethyl-indole
PubChem CID 15427
IUPAC Name 2,3,3-trimethylindole
SMILES CC1=NC2=CC=CC=C2C1(C)C

Specifications

Boiling Point 229°C
Quantity 25 mL
Formula Weight 159.23
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material 2,3,3-Trimethylindolenine
EINECSNumber (5)-0133
TSCA Yes