missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethyl-1-butene 98.0+%, TCI America™

Catalog No. p-7136387
Change view
Click to view available options
Quantity:
1 mL
5 mL
Catalog No. Quantity
T06711ML 1 mL
T06715ML 5 mL
2 options

Catalog No. T06711ML

Supplier: TCI America T06711ML

Only null left

Chemical Identifiers

CAS 594-56-9
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00008850
InChI Key AUYRUAVCWOAHQN-UHFFFAOYSA-N
PubChem CID 11669
IUPAC Name 2,3,3-trimethylbut-1-ene
SMILES CC(=C)C(C)(C)C

Specifications

Color Colorless
Boiling Point 78°C
Quantity 1 mL
UN Number 3295
Synonym 2,3,3-trimethyl-1-butene, 1-butene, 2,3,3-trimethyl, triptene, acmc-209mdj, 2,3-trimethyl-1-butene, 1-butene,3,3-trimethyl, 1-butene,2,3,3-trimethyl, ch3 3cc ch3 =ch2
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,3-Trimethyl-1-butene