missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Thiodiglycolic Acid 98.0+%, TCI America™
Supplier: TCI America T1248500G
Specifications
| 2,2′-Thiodiglycolic Acid | |
| 123-93-3 | |
| C4H6O4S | |
| 3261 | |
| UVZICZIVKIMRNE-UHFFFAOYSA-N | |
| 2-(carboxymethylsulfanyl)acetic acid | |
| 31277 | |
| 150.15 | |
| 500 g |
| 130°C | |
| White | |
| MFCD00004359 | |
| thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid | |
| C(C(=O)O)SCC(=O)O | |
| 150.148 | |
| CHEBI:35012 | |
| ≥98.0% (GC,T) | |
| Crystalline Powder |
Chemical Identifiers
| 123-93-3 | |
| 150.148 | |
| UVZICZIVKIMRNE-UHFFFAOYSA-N | |
| 31277 | |
| 2-(carboxymethylsulfanyl)acetic acid |
| C4H6O4S | |
| MFCD00004359 | |
| thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid | |
| CHEBI:35012 | |
| C(C(=O)O)SCC(=O)O |
Safety and Handling
RTECSNumber : AJ6475000
TSCA : Yes