missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thiodiglycolic Acid 98.0+%, TCI America™

Supplier:  TCI America T124825G

 View more versions of this product

Catalog No. T124825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2′-Thiodiglycolic Acid
123-93-3
C4H6O4S
3261
UVZICZIVKIMRNE-UHFFFAOYSA-N
2-(carboxymethylsulfanyl)acetic acid
31277
150.15
25 g
130°C
White
MFCD00004359
thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
C(C(=O)O)SCC(=O)O
150.148
CHEBI:35012
≥98.0% (GC,T)
Crystalline Powder

Chemical Identifiers

123-93-3
150.148
UVZICZIVKIMRNE-UHFFFAOYSA-N
31277
2-(carboxymethylsulfanyl)acetic acid
C4H6O4S
MFCD00004359
thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
CHEBI:35012
C(C(=O)O)SCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AJ6475000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions