missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)pyridine 98.0+%, TCI America™

Catalog No. T29365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29365G 5 g
T293625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29365G Supplier TCI America Supplier No. T29365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3319-99-1
Molecular Formula C9H7NS
Molecular Weight (g/mol) 161.22
MDL Number MFCD00044441
InChI Key QLPKTAFPRRIFQX-UHFFFAOYSA-N
Synonym 2-2-thienyl pyridine, 2-thiophen-2-yl pyridine, 2-thien-2-ylpyridine, 2-2-pyridyl thiophene, pyridine, 2-2-thienyl, thienyl pyridine, 2-thienylpyridine, 2-pyrid-2-ylthiophene, acmc-1cknd, 2-thien-2-yl pyridine
PubChem CID 76832
IUPAC Name 2-(thiophen-2-yl)pyridine
SMILES S1C=CC=C1C1=CC=CC=N1

Specifications

Melting Point 63°C
Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 161.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Thienyl)pyridine