Learn More
2-(2-Thienyl)ethanol, 98%
CAS: 5402-55-1 | C6H8OS | 128.19 g/mol
$114.73 - $114.73
Chemical Identifiers
| CAS | 5402-55-1 |
|---|---|
| Molecular Formula | C6H8OS |
| Molecular Weight (g/mol) | 128.19 |
| MDL Number | MFCD00005462 |
| InChI Key | VMJOFTHFJMLIKL-UHFFFAOYSA-N |
| Synonym | 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol |
| PubChem CID | 79400 |
| IUPAC Name | 2-thiophen-2-ylethanol |
| SMILES | OCCC1=CC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC366540250
|
Thermo Scientific Chemicals
366540250 |
25 g | Glass bottle |
Each for $114.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5402-55-1 | |
| 128.19 | |
| VMJOFTHFJMLIKL-UHFFFAOYSA-N | |
| 79400 | |
| OCCC1=CC=CS1 |
| C6H8OS | |
| MFCD00005462 | |
| 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol | |
| 2-thiophen-2-ylethanol |
Specifications
| 5402-55-1 | |
| 100.0 | |
| 1.1530g/mL | |
| 101°C | |
| 97.5% min. (GC) | |
| C6H8OS | |
| MFCD00005462 | |
| 17, V,3, 379 | |
| 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol | |
| VMJOFTHFJMLIKL-UHFFFAOYSA-N | |
| 2-thiophen-2-ylethanol | |
| 79400 | |
| 98% | |
| 2-(2-Thienyl)ethanol, 98% |
| 97.5 | |
| Brown or Gray-Green to Yellow | |
| 108°C (13.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5490 to 1.552 | |
| 25 g | |
| 1.153 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in mek,alcohol,organic halogen solvents | |
| OCCC1=CC=CS1 | |
| 128.19 | |
| 128.19 | |
| Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 226-452-
Recommended Storage : this product discolors on storage
RUO – Research Use Only