missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thenil, 98+%

Catalog No. ac393630010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393630010 1 g Glass bottle
AC393630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393630010 Supplier Thermo Scientific Chemicals Supplier No. 393630010
Only null left

CAS: 7333-07-5 | C10H6O2S2 | 222.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7333-07-5
Molecular Formula C10H6O2S2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00173678
InChI Key UNWKVSDABPCZMK-UHFFFAOYSA-N
Synonym 2,2'-thenil, 1,2-di thiophen-2-yl ethane-1,2-dione, bis thiophen-2-yl ethane-1,2-dione, ethanedione, di-2-thienyl, 1,2-di 2-thienyl-1,2-ethanedione, 1,2-di 2-thienyl ethane-1,2-dione, 1,2-di-thiophen-2-yl-ethane-1,2-dione, 5-alpha-thenil, dithienylethanedione, 2,2'-bithenoyl
PubChem CID 275005
IUPAC Name 1,2-dithiophen-2-ylethane-1,2-dione
SMILES C1=CSC(=C1)C(=O)C(=O)C2=CC=CS2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 81°C to 84°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 222.29
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 2,2′-Thenil
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only