Learn More
MilliporeSigma™ Supelco™ 2-(2-Pyridyl)ethyl Silica Gel
Catalog No. 111008971
Change view
Click to view available options
Quantity:
108/Pk.
Catalog No. Quantity
111008971 108/Pk.
1 options
Catalog No. 111008971 Supplier MilliporeSigma™ Supelco™ Supplier No. 54127U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Weak anion exchanger ideal for extracting strong basic compounds that remain charged at all pH levels.
Retention Mechanism: Anion-exchange. Sample Matrix Compatibility: Organic or aqueous Solutions. Weak anion exchanger ideal for extracting strong basic compounds that remain charged at all pH levels. Unlike conventional weak anion exchange SPE phases such as -NH2 (aminopropyl) that have a pKa of 9-10, a pH = 7. This characteristic is important for extracting analytes that are not stable (e.g. hydrolyzes) at high pHs typically required for elution when using traditional weak anion exchangers. Ideal for extracting acyl-coenzyme A esters from tissue.
Specifications
Technique SPE
Quantity 108/Pk.
Description Bed wt. 100 mg, Volume: 1 mL
Material Silica