missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)ethanesulfonic Acid 98.0+%, TCI America™

Catalog No. p-7134569
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03525G 5 g
P035225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03525G Supplier TCI America Supplier No. P03525G

Chemical Identifiers

CAS 68922-18-9
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00006365
InChI Key VRBVUAYEXFCDPE-UHFFFAOYSA-N
PubChem CID 96724
IUPAC Name 2-(pyridin-2-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCC1=CC=CC=N1

Specifications

Melting Point 268°C
Color White-Yellow
Quantity 5 g
Formula Weight 187.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Pyridyl)ethanesulfonic Acid