missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)benzoxazole 97.0+%, TCI America™

Catalog No. P20981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20981G 1 g
P20985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20981G Supplier TCI America Supplier No. P20981G

Chemical Identifiers

CAS 32959-62-9
Molecular Formula C12H8N2O
Molecular Weight (g/mol) 196.21
MDL Number MFCD00569582
InChI Key WELSCYIRWKBEBZ-UHFFFAOYSA-N
PubChem CID 135005
IUPAC Name 2-(pyridin-2-yl)-1,3-benzoxazole
SMILES O1C2=CC=CC=C2N=C1C1=CC=CC=N1

Specifications

Melting Point 103°C
Boiling Point 150°C
Quantity 1 g
Formula Weight 196.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Pyridyl)benzoxazole