missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Propynyloxy)tetrahydro-2H-pyran, 98%

Catalog No. AC295060250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295060250 25 g Glass bottle
AC295061000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295060250 Supplier Thermo Scientific Chemicals Supplier No. 295060250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6089-04-9 | C8H12O2 | 140.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6089-04-9
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00006604
InChI Key HQAXHIGPGBPPFU-UHFFFAOYNA-N
Synonym tetrahydro-2-2-propynyloxy-2h-pyran, 2-2-propynyloxy tetrahydropyran, 2-propargyloxane, 2-prop-2-yn-1-yloxy tetrahydro-2h-pyran, 2-propargyloxy tetrahydropyran, propargyl 2-tetrahydropyranyl ether, propargyl alcohol tetrahydropyranyl ether, 2h-pyran, tetrahydro-2-2-propynyloxy, 2-propynyl 2-pyranyl ether, 3-2'-tetrahydropyranyloxy propyne
PubChem CID 98609

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9970g/mL
Boiling Point 63°C to 65°C (9.0 mmHg)
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.458
Quantity 25 g
Specific Gravity 0.997
Formula Weight 140.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-Propynyloxy)tetrahydro-2H-pyran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-027-5

RUO – Research Use Only