missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Phthalimidoethoxy)acetic acid, 97%

Catalog No. AAH3398806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3398806 5 g
AAH3398803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3398806 Supplier Thermo Scientific Chemicals Supplier No. H3398806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69676-65-9 | C12H11NO5 | 249.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69676-65-9
Molecular Formula C12H11NO5
Molecular Weight (g/mol) 249.22
MDL Number MFCD06661364
InChI Key PHZYUQLIZKTSJE-UHFFFAOYSA-N
Synonym 2-2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-phthalimidoethoxy acetic acid, 2-2-phthalimidoethoxy-acetic acid, 2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-phthalimido ethoxyacetic acid, 2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindolin-2-yl ethoxy aceticacid, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethoxy acetic acid
PubChem CID 2755407
IUPAC Name 2-[2-(1,3-dioxoisoindol-2-yl)ethoxy]acetic acid
SMILES OC(=O)COCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 130°C to 132°C
Quantity 5 g
Formula Weight 249.22
Percent Purity 97%
Chemical Name or Material 2-(2-Phthalimidoethoxy)acetic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only