missing translation for 'onlineSavingsMsg'
Learn More

[2.2]Paracyclophane, 99%

CAS: 1633-22-3 | C16H16 | 208.304 g/mol

$110.86 - $354.82

Chemical Identifiers

CAS 1633-22-3
Molecular Formula C16H16
Molecular Weight (g/mol) 208.304
MDL Number MFCD00003707
InChI Key OOLUVSIJOMLOCB-UHFFFAOYSA-N
Synonym 2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
PubChem CID 74210
SMILES C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2013603
View Documents
Thermo Scientific Chemicals
B2013603
1 g
Each for $110.86
Only null left
 
AAB2013606
View Documents
Thermo Scientific Chemicals
B2013606
5 g
Each for $354.82
Only null left
 
Description

Description

2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1633-22-3
208.304
OOLUVSIJOMLOCB-UHFFFAOYSA-N
74210
C16H16
MFCD00003707
2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2

Specifications

1633-22-3
Odorless
MFCD00003707
1910888
Insoluble in water.
C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
74210
99%
283°C to 288°C
C16H16
1 g
2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
OOLUVSIJOMLOCB-UHFFFAOYSA-N
208.304
208.3
[2.2]Paracyclophane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-644-2

TSCA : Yes

SDS
Documents

Documents

RUO – Research Use Only