missing translation for 'onlineSavingsMsg'
Learn More

[2,2]-Paracyclophane, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 291600010

 View more versions of this product

Catalog No. AC291600010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

[2,2]-Paracyclophane
Authentic
Glass bottle
1g
Solubility in water: insoluble. Other solubilities: soluble in toluene
C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
74210
99%
1633-22-3
98.5% min. (HPLC)
C16H16
2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
OOLUVSIJOMLOCB-UHFFFAOYSA-N
208.3
208.3

Chemical Identifiers

1633-22-3
208.3
2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
C16H16
OOLUVSIJOMLOCB-UHFFFAOYSA-N
74210
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-644-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only