missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Oxybis(ethylamine) 98.0+%, TCI America™
Supplier: TCI America O027825G
Specifications
| 2,2′-Oxybis(ethylamine) | |
| Yellow | |
| C4H12N2O | |
| 25 g | |
| 2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine | |
| NCCOCCN | |
| 104.15 | |
| 104.15 | |
| Liquid |
| 2752-17-2 | |
| 64°C | |
| MFCD00466591 | |
| 2735 | |
| GXVUZYLYWKWJIM-UHFFFAOYSA-N | |
| 2-(2-aminoethoxy)ethan-1-amine | |
| 75982 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 2752-17-2 | |
| 104.15 | |
| GXVUZYLYWKWJIM-UHFFFAOYSA-N | |
| 75982 | |
| NCCOCCN |
| C4H12N2O | |
| MFCD00466591 | |
| 2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine | |
| 2-(2-aminoethoxy)ethan-1-amine |
Safety and Handling
TSCA : Yes