missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Oxybis(ethylamine) 98.0+%, TCI America™

Catalog No. O02781G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02781G 1 g
O02785G 5 g
O027825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O02781G Supplier TCI America Supplier No. O02781G

Chemical Identifiers

CAS 2752-17-2
Molecular Formula C4H12N2O
Molecular Weight (g/mol) 104.15
MDL Number MFCD00466591
InChI Key GXVUZYLYWKWJIM-UHFFFAOYSA-N
Synonym 2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine
PubChem CID 75982
IUPAC Name 2-(2-aminoethoxy)ethan-1-amine
SMILES NCCOCCN

Specifications

Color Yellow
Boiling Point 64°C
Quantity 1 g
UN Number 2735
Formula Weight 104.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2′-Oxybis(ethylamine)