missing translation for 'onlineSavingsMsg'
Learn More

2,2'-O-Cyclouridine 98.0+%, TCI America™

Catalog No. C22085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22085G 5 g
C220825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22085G Supplier TCI America Supplier No. C22085G

Chemical Identifiers

CAS 3736-77-4
Molecular Formula C9H10N2O5
Molecular Weight (g/mol) 226.188
MDL Number MFCD00004945
InChI Key UUGITDASWNOAGG-CCXZUQQUSA-N
Synonym cyclouridine, 2,2'-cyclouridine, o2,2'-cyclouridine, 2,2'-anhydro-1 b-d-arabinofuranosyl uracil, 2,2'-anhydrouridine, 2,2'-anhydro-1 beta-d-arabinofuranosyl uracil, 2,2'-o-cyclouridine, 2,2-anhydro-1 b-d-arabinofuranosyl uracil, 2,2;-cyclouridine, 2,2'-anhydro-1-beta-d-arabinofuranosyl uracil
PubChem CID 806138
IUPAC Name (2R,3R,3aS,9aR)-3-hydroxy-2-(hydroxymethyl)-2,3,3a,9a-tetrahydrofuro[1,2][1,3]oxazolo[3,4-a]pyrimidin-6-one
SMILES C1=CN2C3C(C(C(O3)CO)O)OC2=NC1=O

Specifications

Melting Point 244°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 226.19
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,2′-O-Cyclouridine
TSCA No
Recommended Storage Freezer