missing translation for 'onlineSavingsMsg'
Learn More

2,2'-O-Cyclouridine, 98%

Catalog No. AAH3303803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3303803 1 g
AAH3303806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3303803 Supplier Thermo Scientific Chemicals Supplier No. H3303803
Only null left

CAS: 3736-77-4 | C9H10N2O5 | 226.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3736-77-4
Molecular Formula C9H10N2O5
Molecular Weight (g/mol) 226.188
MDL Number MFCD00004945
InChI Key UUGITDASWNOAGG-CCXZUQQUSA-N
Synonym cyclouridine, 2,2'-cyclouridine, o2,2'-cyclouridine, 2,2'-anhydro-1 b-d-arabinofuranosyl uracil, 2,2'-anhydrouridine, 2,2'-anhydro-1 beta-d-arabinofuranosyl uracil, 2,2'-o-cyclouridine, 2,2-anhydro-1 b-d-arabinofuranosyl uracil, 2,2;-cyclouridine, 2,2'-anhydro-1-beta-d-arabinofuranosyl uracil
PubChem CID 806138
IUPAC Name (2R,3R,3aS,9aR)-3-hydroxy-2-(hydroxymethyl)-2,3,3a,9a-tetrahydrofuro[1,2][1,3]oxazolo[3,4-a]pyrimidin-6-one
SMILES C1=CN2C3C(C(C(O3)CO)O)OC2=NC1=O

Specifications

Melting Point 248°C to 251°C
Quantity 1 g
Beilstein 27348885
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material 2,2'-O-Cyclouridine
EINECSNumber 223-107-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only