missing translation for 'onlineSavingsMsg'
Learn More

2,2'-O-Cyclocytidine Hydrochloride 98.0+%, TCI America™

Catalog No. C22071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22071G 1 g
C22075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22071G Supplier TCI America Supplier No. C22071G

Chemical Identifiers

CAS 10212-25-6
Molecular Formula C9H12ClN3O4
Molecular Weight (g/mol) 261.66
MDL Number MFCD00012636
InChI Key KZOWNALBTMILAP-JBMRGDGGSA-N
Synonym cyclocytidine hydrochloride 1g
PubChem CID 74764394
ChEBI CHEBI:74843
IUPAC Name (2R,4R,5R,6S)-4-(hydroxymethyl)-10-imino-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-5-ol hydrochloride
SMILES Cl.OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O

Specifications

Melting Point 264°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-O-Cyclocytidine Hydrochloride
RTECSNumber LV2615000
TSCA No