missing translation for 'onlineSavingsMsg'
Learn More

2-(2-n-Butoxyethoxy)ethyl acetate, 98%

Catalog No. AAA16065AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16065AU 1000 mL
AAA16065AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16065AU Supplier Thermo Scientific Chemicals Supplier No. A16065AU

CAS: 124-17-4 | C10H20O4 | 204.266 g/mol

2-(2-n-Butoxyethoxy)ethyl acetate is widely used in high-grade paints for car, TV, freezer and plane. It is used as a solvent for enhancement of coating's characters in the coating industry. It is also used as a cleaning and furnishing care product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-n-Butoxyethoxy)ethyl acetate is widely used in high-grade paints for car, TV, freezer and plane. It is used as a solvent for enhancement of coating′s characters in the coating industry. It is also used as a cleaning and furnishing care product.

Solubility
Miscible with water, acetone, ethyl ether, ethanol and most organic liquids.

Notes
Light sensitive and heat sensitive. Incompatible with strong oxidizing agents, acids and strong bases. Keep the container tightly closed.

Chemical Identifiers

CAS 124-17-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00009458
InChI Key VXQBJTKSVGFQOL-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethyl acetate, butoxyethoxyethyl acetate, diethylene glycol monobutyl ether acetate, butyl diglycol acetate, butyl carbitol acetate, ektasolve db acetate, 2-2-butoxyethoxy ethanol acetate, butyl diethylene glycol acetate, butylkarbitolacetat, diglycol monobutyl ether acetate
PubChem CID 31288
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
SMILES CCCCOCCOCCOC(=O)C

Specifications

Melting Point °C to 32°C
Density 0.977
Boiling Point 237°C to 241°C
Flash Point 115°C (239°F)
Assay Percent Range 98%
Refractive Index 1.426
Quantity 1000 mL
Beilstein 1771533
Solubility Information Miscible with water,acetone,ethyl ether,ethanol and most organic liquids.
Formula Weight 204.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-n-Butoxyethoxy)ethyl Acetate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
EINECSNumber 204-685-9
RTECSNumber KJ9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only