missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1H-imidazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC61216DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC61216DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

892502-27-1
183.214
NCQQCFZREDABTC-UHFFFAOYSA-N
24229637
CC1=NC=CN1C2=CC=CC=C2C#N
C11H9N3
MFCD09065013
2-2-methyl-1h-imidazol-1-yl benzonitrile, 2-2-methylimidazol-1-yl benzonitrile, 2-2-methylimidazolyl benzenecarbonitrile, benzonitrile,2-2-methyl-1h-imidazol-1-yl
2-(2-methylimidazol-1-yl)benzonitrile

Specifications

2-(2-Methyl-1H-imidazol-1-yl)benzonitrile
96.5
97%
C11H9N3
MFCD09065013
NCQQCFZREDABTC-UHFFFAOYSA-N
2-(2-methylimidazol-1-yl)benzonitrile
24229637
97%
892502-27-1
100.0
Amber Glass Bottle
5g
2-2-methyl-1h-imidazol-1-yl benzonitrile, 2-2-methylimidazol-1-yl benzonitrile, 2-2-methylimidazolyl benzenecarbonitrile, benzonitrile,2-2-methyl-1h-imidazol-1-yl
CC1=NC=CN1C2=CC=CC=C2C#N
183.214
183.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions