missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)ethanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC54609DA

Quantity 1g

 View more versions of this product

Catalog No. CC54609DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

121357-04-8
143.204
2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole
2-(2-methyl-1,3-thiazol-4-yl)ethanol
C6H9NOS
REKVZAFNJAMAQL-UHFFFAOYSA-N
19876739
CC1=NC(=CS1)CCO

Specifications

2-(2-Methyl-1, 3-thiazol-4-yl)ethanol
97%
1g
REKVZAFNJAMAQL-UHFFFAOYSA-N
2-(2-methyl-1,3-thiazol-4-yl)ethanol
19876739
121357-04-8
C6H9NOS
2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole
CC1=NC(=CS1)CCO
143.204
143.21
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents