missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Methoxyethoxy)ethoxy]acetic Acid 95.0+%, TCI America™

Catalog No. p-7133515
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29635G 5 g
M296325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29635G Supplier TCI America Supplier No. M29635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16024-58-1
Molecular Formula C7H14O5
Molecular Weight (g/mol) 178.184
MDL Number MFCD00044098
InChI Key YHBWXWLDOKIVCJ-UHFFFAOYSA-N
Synonym 3,6,9-Trioxadecanoic Acid
PubChem CID 85241
IUPAC Name 2-[2-(2-methoxyethoxy)ethoxy]acetic acid
SMILES COCCOCCOCC(=O)O

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
Formula Weight 178.18
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material [2-(2-Methoxyethoxy)ethoxy]acetic Acid