missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxyphenyl)benzoxazole 98.0+%, TCI America™
Supplier: TCI America H029125G
Specifications
| 2-(2-Hydroxyphenyl)benzoxazole | |
| 124°C | |
| C13H9NO2 | |
| 25 g | |
| UWUSYRFTCSSHAJ-UHFFFAOYSA-N | |
| 6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one | |
| 6744797 | |
| ≥98.0% (GC,T) |
| 835-64-3 | |
| 338°C | |
| MFCD00005767 | |
| 2-2-hydroxyphenyl benzoxazole, 6-3h-1,3-benzoxazol-2-ylidene cyclohexa-2,4-dien-1-one, 2-2-benzoxazolyl phenol, ghgzvwotjdlrey-uhfffaoysa-n, phenol, benzoxazolyl, acmc-209psp, 2-2'-hydroxyphenyl benzoxazole, 2,4-cyclohexadien-1-one, 6-2 3h-benzoxazolylidene | |
| C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2 | |
| 211.22 | |
| 211.22 | |
| Crystalline Powder |
Chemical Identifiers
| 835-64-3 | |
| 211.22 | |
| UWUSYRFTCSSHAJ-UHFFFAOYSA-N | |
| 6744797 | |
| C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2 |
| C13H9NO2 | |
| MFCD00005767 | |
| 2-2-hydroxyphenyl benzoxazole, 6-3h-1,3-benzoxazol-2-ylidene cyclohexa-2,4-dien-1-one, 2-2-benzoxazolyl phenol, ghgzvwotjdlrey-uhfffaoysa-n, phenol, benzoxazolyl, acmc-209psp, 2-2'-hydroxyphenyl benzoxazole, 2,4-cyclohexadien-1-one, 6-2 3h-benzoxazolylidene | |
| 6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one |
Safety and Handling
RTECSNumber : SJ7520000
TSCA : Yes