missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyphenyl)benzoxazole 98.0+%, TCI America™

Catalog No. H02911G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02911G 1 g
H029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02911G Supplier TCI America Supplier No. H02911G

Chemical Identifiers

CAS 835-64-3
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.22
MDL Number MFCD00005767
InChI Key UWUSYRFTCSSHAJ-UHFFFAOYSA-N
Synonym 2-2-hydroxyphenyl benzoxazole, 6-3h-1,3-benzoxazol-2-ylidene cyclohexa-2,4-dien-1-one, 2-2-benzoxazolyl phenol, ghgzvwotjdlrey-uhfffaoysa-n, phenol, benzoxazolyl, acmc-209psp, 2-2'-hydroxyphenyl benzoxazole, 2,4-cyclohexadien-1-one, 6-2 3h-benzoxazolylidene
PubChem CID 6744797
IUPAC Name 6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2

Specifications

Melting Point 124°C
Boiling Point 338°C
Quantity 1 g
Formula Weight 211.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Hydroxyphenyl)benzoxazole
RTECSNumber SJ7520000
TSCA Yes