missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyphenyl)benzothiazole 98.0+%, TCI America™

Catalog No. H09735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09735G 5 g
H097325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09735G Supplier TCI America Supplier No. H09735G

Chemical Identifiers

CAS 3411-95-8
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00022869
InChI Key NSDGEQWRYXOZLN-UKTHLTGXSA-N
Synonym phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
PubChem CID 5376551
IUPAC Name 6-[(2E)-2,3-dihydro-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 227.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Hydroxyphenyl)benzothiazole
RTECSNumber SJ7360000
TSCA Yes