missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyphenyl)benzothiazole, 98%

Catalog No. AAA1934614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1934614 25 g
AAA1934606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1934614 Supplier Thermo Scientific Chemicals Supplier No. A1934614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3411-95-8 | C13H9NOS | 227.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3411-95-8
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00022869
InChI Key NSDGEQWRYXOZLN-UKTHLTGXSA-N
Synonym phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
PubChem CID 5376551
SMILES O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1

Specifications

Melting Point 130°C to 134°C
Quantity 25 g
Beilstein 173026
IUPAC Name (6Z)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one
Formula Weight 227.29
Percent Purity 98%
Chemical Name or Material 2-(2-Hydroxyphenyl)benzothiazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-299-9
RTECSNumber SJ7360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only