missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxyethoxy)benzaldehyde 96.0+%, TCI America™
$167.76 - $458.56
Chemical Identifiers
| CAS | 22042-72-4 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00191449 |
| InChI Key | WGHPWLUYIPUQOJ-UHFFFAOYSA-N |
| PubChem CID | 93989 |
| IUPAC Name | 2-(2-hydroxyethoxy)benzaldehyde |
| SMILES | OCCOC1=CC=CC=C1C=O |
Chemical Identifiers
| 22042-72-4 | |
| 166.18 | |
| WGHPWLUYIPUQOJ-UHFFFAOYSA-N | |
| 2-(2-hydroxyethoxy)benzaldehyde |
| C9H10O3 | |
| MFCD00191449 | |
| 93989 | |
| OCCOC1=CC=CC=C1C=O |
Specifications
| 22042-72-4 | |
| 134°C | |
| MFCD00191449 | |
| WGHPWLUYIPUQOJ-UHFFFAOYSA-N | |
| 2-(2-hydroxyethoxy)benzaldehyde | |
| 93989 | |
| ≥96.0% (GC) | |
| 2-(2-Hydroxyethoxy)benzaldehyde |
| 45°C | |
| C9H10O3 | |
| 5 g | |
| OCCOC1=CC=CC=C1C=O | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator