missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyethoxy)benzaldehyde 96.0+%, TCI America™

Catalog No. H08575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08575G 5 g
H085725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08575G Supplier TCI America Supplier No. H08575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22042-72-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00191449
InChI Key WGHPWLUYIPUQOJ-UHFFFAOYSA-N
PubChem CID 93989
IUPAC Name 2-(2-hydroxyethoxy)benzaldehyde
SMILES OCCOC1=CC=CC=C1C=O

Specifications

Melting Point 45°C
Boiling Point 134°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Hydroxyethoxy)benzaldehyde
TSCA No
Recommended Storage Refrigerator