missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Furyl)phenyl]methylamine, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC46213DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC46213DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

771573-25-2
173.215
KUTREPQQCILHIM-UHFFFAOYSA-N
18525791
C1=CC=C(C(=C1)CN)C2=CC=CO2
C11H11NO
MFCD06213664
2-2-furyl phenyl methylamine, 2-furan-2-yl phenyl methanamine, 1-2-furan-2-yl phenyl methanamine, 2-furan-2-yl-benzylamine
[2-(furan-2-yl)phenyl]methanamine

Specifications

[2-(2-Furyl)phenyl]methylamine
96.5
97%
C11H11NO
MFCD06213664
KUTREPQQCILHIM-UHFFFAOYSA-N
[2-(furan-2-yl)phenyl]methanamine
18525791
97%
771573-25-2
100.0
Amber Glass Bottle
1g
2-2-furyl phenyl methylamine, 2-furan-2-yl phenyl methanamine, 1-2-furan-2-yl phenyl methanamine, 2-furan-2-yl-benzylamine
C1=CC=C(C(=C1)CN)C2=CC=CO2
173.215
173.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions