missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Furyl)benzonitrile 97+%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 169.18 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(2-Furyl)benzonitrile |
Safety and Handling
| Hazard Category | Warning |