missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. p-4510689
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46201CB 250mg Amber Glass Bottle
CC46201DA 1g Amber Glass Bottle
CC46201DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC46201CB Supplier Thermo Scientific Chemicals Supplier No. CC46201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 331942-47-3
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
InChI Key QRUHYAWZHFTNEA-UHFFFAOYSA-N
Synonym 2-2-furyl benzoic acid, 2-furan-2-yl benzoic acid, 2-fur-2-ylbenzoic acid, benzoic acid,2-2-furanyl, 2-fur-2-yl benzoic acid, 2-furan-2-yl-benzoic acid
PubChem CID 2772293
IUPAC Name 2-(furan-2-yl)benzoic acid
SMILES C1=CC=C(C(=C1)C2=CC=CO2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material 2-(2-Furyl)benzoic acid