missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Furoin, 98%

Catalog No. AAA1527206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1527206 5 g
AAA1527214 25 g
AAA1527222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1527206 Supplier Thermo Scientific Chemicals Supplier No. A1527206

CAS: 552-86-3 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-86-3
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003246
InChI Key MIJRFWVFNKQQDK-UHFFFAOYNA-N
Synonym furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone
PubChem CID 11100
IUPAC Name 1,2-bis(furan-2-yl)-2-hydroxyethanone
SMILES OC(C(=O)C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 137°C to 139°C
Quantity 5 g
Beilstein 383995
Formula Weight 192.17
Percent Purity 98%
Chemical Name or Material 2,2'-Furoin
EINECSNumber 209-024-8
RTECSNumber KM5774095
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only